2-Chlor-3-methylpyrazin

Lieferant: BEPHARM

B2664-5G
BEPHB2664-5GEA 0 EUR
BEPHB2664-5G
2-Chlor-3-methylpyrazin
2-Chloro-3-methylpyrazine
Formel: C₅H₅ClN₂
Molecular Weight: 128,56 g/mol
MDL: MFCD00023279
CAS-Nummer: 95-58-9
EINECS: 202-434-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR