4-Brom-N-phenylanilin

Lieferant: BEPHARM

B95236-100G
BEPHB95236-100GEA 0 EUR
BEPHB95236-100G
4-Brom-N-phenylanilin
4-Brom-N-phenylanilin
Formel: C₁₂H₁₀BrN
Molecular Weight: 248,1200 g/mol
MDL: MFCD07779504
CAS-Nummer: 54446-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR