Mop head frame, small, flat, BCR® Mop 3
Lieferant: BERKSHIRE
BCR.MOP3.1
BERKBCR.MOP3.1EA 429 EUR
BERKBCR.MOP3.1
Mop head frame, small, flat, BCR® Mop 3
Mopps
Berkshire’s flat mop head is light and easy to maneuver, even with wet wipers attached. The stainless steel springs or plastic eyes hold multiple mop wipers.
  • Clean, light, and durable
  • High quality anodized aluminum
  • Autoclavable (plastic eyes are not autoclavable)
  • Plastic eyes for wiper attachment
Efficient cleaning of floors, walls, and other large surfaces is critical to the overall cleanliness of the production environment. BCR® mop systems replicate the most effective way to clean a surface wiping. Berkshire’s mop systems are easy to use, highly effective, and quick.
The system is very flexible, combining a wide range of alternative mop wipers designed for particular levels of cleanliness with a range of flat mop heads.
Standards: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.