SatPax® Polx® Pre-wetted, 100% Polyester Non-woven Cleanroom Wipes, Canister and Canister Refills
Lieferant: BERKSHIRE
SPXCPNW00412
BERKSPXCPNW00412EA 924 EUR
BERKSPXCPNW00412
SatPax® Polx® Pre-wetted, 100% Polyester Non-woven Cleanroom Wipes, Canister and Canister Refills
Wischtücher Reinraum-Wischtücher Getränkte Reinraumwischtücher
SatPax® Polx® non-woven combines Polx non-woven wipers, composed of 100% polyester spunlace, with various saturation levels of isopropyl alcohol and DI water in an easy-to-dispense canister.
  • Exceptional cleanliness
  • Prevents abrasion with an extremely soft and pliable surface
  • Increases cleaning protocol consistency
  • Lot to lot traceability
  • Reduces alcohol usage and preparation/handling costs
  • Increases cleaning efficiency
In addition to being a cost-effective and easy-to-use solution, this prewetted format of Polx® non-woven has critically low fiber and particle generation and minimal non-volatile residues, making it an exceptional choice for streak-free cleaning.
Wipers are available in three concentration formats; pre-wetted with 70% IPA, 30% DI water, 60% saturation, pre-wetted with 9% IPA, 91% DI water, 60% saturation, or pre-wetted with 99% IPA, 1% DI water, 60% saturation.
Standards: ISO Class 5 to 8. Cleanroom Class 100 to 100,000. EU Grade B/C/D.
Verpackung: Wipers are packaged 100 sheets per pack, 12 packs per case, for a total of 1200 sheets per case.