Cleanroom wipes, sterile, sealed edge, ValuSeal® IonX™
Lieferant: BERKSHIRE
VSI.0909.14
BERKVSI.0909.14EA 1370 EUR
BERKVSI.0909.14
Cleanroom wipes, sterile, sealed edge, ValuSeal® IonX™
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
Designed for the highest level of contamination control in critical processing applications.
  • Critically low particles, fibres, ions, and extractables
  • High abrasion resistance
  • Lightweight, high absorbency material reduces actual cost in use compared to heavier weight products
  • Laser sealed edge for reduced fibre contamination
ValuSeal® IonX® is a laser sealed edge cleanroom laundered wipe recommended for ISO Class 3 and above environments. Composed of 100% polyester continuous filament yarns, its smooth knit construction and laser sealed edges make it ideal for applications requiring critically low levels of particle and fibre release, along with durability and chemical compatibility.
Standards: ISO Class 3 – 5. Cleanroom Class 1 – 100. EU Grade B/C/D.