Sealing ring for reaction vessels
Lieferant: SciLabware
296-90
BIBB296-90EA 66.4 EUR
BIBB296-90
Sealing ring for reaction vessels
Versiegeln Dichtringe
Silicone rubber sheathed with PTFE.
  • Guarantee excellent joint seal
  • Temperature resistant from −200 to +200 °C
  • Excellent chemical resistance
O-rings for flat flanges with groove.