Burettes, straight stopcock, MBL®
Lieferant: SciLabware
BRB070P BRB074P BRB050P BRB056P BRB058P BRB060P BRB076P BRB075P BRB077P
BIBBBRB070PEA 36.2 EUR
BIBBBRB070P BIBBBRB074P 612-0351 612-0352 612-0353 612-0354 BIBBBRB076P BIBBBRB075P BIBBBRB077P
Burettes, straight stopcock, MBL®
Büretten
Borosilicate glass with interchangeable key, class B.
  • Graduations in easy to read blue enamel
  • Key/stopcock with 1,5 mm bore
  • Choice of glass key or grease-free PTFE stopcock
* burettes fitted with funnel tops to make filling easier.
Standards: DIN EN ISO 385