O-ring seals for flat flanges, Quickfit®
Lieferant: SciLabware
PS100 PS75
BIBBPS100EA 31.6 EUR
BIBBPS100 BIBBPS75
O-ring seals for flat flanges, Quickfit®
Versiegeln Dichtringe
PTFE, white.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Forms tight seal with flat flange joints
For use with Quickfit® flat flange jointed glassware.