The All-In-One Highline is the design concept that has an especially high back. It has a stable tubular steel base with flat lines. All the steel parts are painted black. The plastic parts are black too, and extremely tough. The high version features a chromed foot ring that is height-adjustable, and can be used as foot support and as a mounting aid. The chair can be fitted either with castors with load-sensitive brakes for hard floors, or with abrasion-resistant glides. On request, the high version can also be supplied with Stop and Go castors.
- Synchronous mechanism with weight adjustment
- Ergonomic package composed of:
- Adjustable seat depth and seat inclination
- Backrest adjustable in height
- Polyamide base
A real all-rounder - the All-In-One is closing the gap between office and work chairs. All the ergonomic properties expected of an office chair can be found in the Highline and Trend design lines. Its unique all-inclusive ergonomic concept ensures that the chair suits every single person and every single workplace situation. As a work chair for industrial sectors, healthcare, research and development, it boasts a sturdy construction and hard-wearing, resistant materials.
Seat finish: Duotec fabric, Skai synthetic leather, Integral foam.
Standards: DIN 68 877, German GS 'safety tested' mark of confirmity, Quality Management DIN EN ISO 9001, Environmental Management DIN ISO 14001, Certified Environmental Management EMAS, Occupational Safety Management OHSAS 18001, EcoVadis CSR rating 2015 Gold.