Analogue stopwatch

Lieferant: Binhold, Paul Lehrmittel
1002809
BINH1002809EA 372 EUR
BINH1002809
Analogue stopwatch
Timer Stoppuhren, analog
Large, quartz controlled stopwatch for benchtop use.

  • Start / Stop / Reset buttons
  • Timing capacity: 60 min/ 60 s
  • Resolution: 1 s or 1/100 min
  • Dial with dual scale for minutes and seconds
  • Functions: Cumulative time and lap-time settings
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR