Vane wheel for heat/air flow
Lieferant: 3B Scientific
1003101
BINH1003101EA 0 EUR
BINH1003101
Vane wheel for heat/air flow
Lehrmittel Physik-Lehrmittel Heat/Thermodynamics Learning Activities
Vane wheel device is used for demonstrating air and heat flows generated by a burning candle, water vapour or other heat sources. Made of Aluminium and mountable on a long needle.