Mitosis model
Lieferant: 3B Scientific
1013868
BINH1013868EA 939 EUR
BINH1013868
Mitosis model
Modelle Molekularbiologie
This model series shows the following nine phases of mitosis on the basis of a typical mammal cell at an enlargement of 10000 times. The three-dimensional relief models are painted according to the usual colouring methods of microscopy, making the process of cell division easy to understand. The cell organelles are shown as if opened up in the lower part of the models.
  • 1. Interphase
  • 2. Prophase
  • 3. Early prometaphase
  • 4. Later prometaphase
  • 5. Metaphase
  • 6. Early anaphase
  • 7. Later anaphase
  • 8. Telophase
  • 9. Cytokinesis
Equipped with magnets at the back for teaching purposes, they can be easily arranged on a magnetic board in the classroom.
Bestellinformation: Includes detailed description and handouts for your lessons.
Lieferumfang: Supplied in a storage system (400×600 mm), which can be fastened to the wall.