Trolley track with two trolleys
Lieferant: 3B Scientific
U35000
BINHU35000EA 0 EUR
BINHU35000
Trolley track with two trolleys
Lehrmittel Physik-Lehrmittel Mechanics Learning Activities
Track with two trolleys and other accessories for investigating linear motion. The trolleys move with minimum friction on wheels with high-quality ball-bearings. They are fitted with magnets at their front ends for experiments involving both elastic and inelastic collisions.
  • Regular movements and movements with uniform acceleration
  • Newton’s laws of motion
  • Conservation of energy
  • Elastic and inelastic collisions
It can be used in combination with a photo gate for recording the motion of a trolley.
Applications include:
Lieferumfang: Set includes track (1.8 m) with distance scale, two-point support, single point support with end stop, 1 × trolley (500 g), 1 × trolley with interchangeable buffer pad, additional weight (500 g), 2 × holder for light barrier, pulley and holder, clamp for stand rods, set of contact-breakers and a set of magnets.