Immersion heater
Lieferant: 3B Scientific
U8624110-230
BINHU8624110-230EA 50 EUR
BINHU8624110-230
Immersion heater
Heizgeräte Tauchsieder
Immersion heater with overheating protection (VDE-compliant). Important: operates solely on a mains voltage of 230 V.