Hand dynamometer, Smedley spring-type, Baseline®
Lieferant: Binhold, Paul Lehrmittel
W54654
BINHW54654EA 367 EUR
BINHW54654
Hand dynamometer, Smedley spring-type, Baseline®
Dynamometer
Digital, lightweight Smedley spring dynamometer offers accurate grip strength readout.
  • 5-preset grip sizes
  • Captures and displays maximum grip force in both kilogrammes and pounds
  • Unit assesses results according to age and gender
  • Unit saves/stores results for fast retrieval with up to 19 users
  • The adjustable handle comfortably accommodates any hand size.
Lieferumfang: Supplied with protective carrying case.