Lens cleaning tissues
Lieferant: Ahlstrom-Munksjö
146099 146100 146102 146103 146104
BINZ146099EA 2.8 EUR
BINZ146099 BINZ146100 BINZ146102 BINZ146103 BINZ146104
Lens cleaning tissues
Wischtücher Wisch- und Handtücher, Einweg Linsenreinigungstücher
These very fine lens cleaning tissues are made of pure manila vegetable fibre.
  • Non abrasive
Lint-free paper for cleaning of optical surfaces such as lenses and mirrors of microscopes, etc.
12 g/m²
Bestellen
TECHNISCHE DATEN
Environmentally Preferable