120mm Dmr., Pkg. Mit 100 Filtern Art.-Nr. 3.206.120 Basis Weight g/m² 84 Filtration Speed s / 10 ml Burst Strength, wet kPa ?30 Typical Retention µm 12-15 1 * 1 ST
Lieferant: Ahlstrom-Munksjö
120mm Dmr., Pkg. Mit 100 Filtern Art.-Nr. 3.206.120 Basis Weight g/m² 84 Filtration Speed s / 10 ml Burst Strength, wet kPa ?30 Typical Retention µm 12-15 1 * 1 ST