Real-Time PCR detection kits, Allergen
Lieferant: BIOPREMIER
FS-0049
FS-0050
FS-0051
FS-0052
FS-0053
FS-0054
FS-0055
FS-SP0049
FS-SP0050
FS-SP0051
FS-SP0052
FS-SP0053
FS-SP0054
FS-SP0055
FS-0060
FS-0061
FS-0062
FS-0063
FS-SP0060
FS-SP0061
FS-SP0062
FS-SP0063
BIOPFS-0049EA
574
EUR
BIOPFS-0049
BIOPFS-0050
BIOPFS-0051
BIOPFS-0052
BIOPFS-0053
BIOPFS-0054
BIOPFS-0055
BIOPFS-SP0049
BIOPFS-SP0050
BIOPFS-SP0051
BIOPFS-SP0052
BIOPFS-SP0053
BIOPFS-SP0054
BIOPFS-SP0055
BIOPFS-0060
BIOPFS-0061
BIOPFS-0062
BIOPFS-0063
BIOPFS-SP0060
BIOPFS-SP0061
BIOPFS-SP0062
BIOPFS-SP0063
Real-Time PCR detection kits, Allergen
Mikrobiologische Testsysteme
PCR-Tests
These kits allow the detection of allergic substances. Ideal for controlling and monitoring the various steps of food production.