Europium(III)fluorid

Lieferant: BIONET RESEARCH

003269-0005
BIOT003269-0005EA 0 EUR
BIOT003269-0005
Europium(III)fluorid
Europium(III)fluorid
Formel: EuF₃
Molecular Weight: 208,96 g/mol
Siedepunkt: 2280 °C (1013 hPa)
Schmelzpunkt: 1249,85…1265.85 °C
MDL: MFCD00010998
CAS-Nummer: 13765-25-8
EINECS: 237-368-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR