Gadolinium(III)fluorid

Lieferant: BIONET RESEARCH

Synonyme: Gadoliniumtrifluorid

003805-0010
BIOT003805-0010EA 0 EUR
BIOT003805-0010
Gadolinium(III)fluorid
Gadolinium(III)fluorid
Formel: F₃Gd
Molecular Weight: 214,25 g/mol
Schmelzpunkt: 1231 °C
MDL: MFCD00016113
CAS-Nummer: 13765-26-9
EINECS: 237-369-4
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR