Lutetiumtrifluorid

Lieferant: BIONET RESEARCH

Synonyme: Lutetium(III)fluorid

004858-0005
BIOT004858-0005EA 0 EUR
BIOT004858-0005
Lutetiumtrifluorid
Lutetiumtrifluorid
Formel: LuF₃
Molecular Weight: 231.96 g/mol
Siedepunkt: 2200
Schmelzpunkt: 1182
Dichte: 8.29 g/cm³ (20 °C)
MDL: MFCD00016191
CAS-Nummer: 13760-81-1
EINECS: 237-355-8
UN: 3288
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR