Molekularsieb 3A (0,3 nm, 3 Å)

Lieferant: BIONET RESEARCH

005546-1000
BIOT005546-1000EA 0 EUR
BIOT005546-1000
Molekularsieb 3A (0,3 nm, 3 Å)
Molekularsieb 3A (0,3 nm, 3 Å)
Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL: MFCD00131613
CAS-Nummer: 308080-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR