Siliciumnitrid

Lieferant: BIONET RESEARCH

Synonyme: Silicon(IV) nitride

006960-0500
BIOT006960-0500EA 0 EUR
BIOT006960-0500
Siliciumnitrid
Siliciumnitrid
Formel: N₄Si₃
Molecular Weight: 140.28 g/mol
MDL: MFCD00011230
CAS-Nummer: 12033-89-5
EINECS: 234-796-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR