Yttrium(III)nitrat Hydrat

Lieferant: BIONET RESEARCH
Gefahr

Synonyme: Yttrium trinitrate hydrate

008326-50G
BIOT008326-50GEA 0 EUR
BIOT008326-50G
Yttrium(III)nitrat Hydrat
Yttrium(III)nitrat Hydrat
Formel: N₃O₉Y·H₂O
Molecular Weight: 274,92 g/mol (anhy)
MDL: MFCD00011469
CAS-Nummer: 140191-40-8
EINECS: 233-802-6
UN: 1477
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR