Arsen, pieces 2-12 mm

Lieferant: BIONET RESEARCH
Gefahr

Synonyme: Arsenic solution

009029
BIOT009029EA 0 EUR
BIOT009029
Arsen, pieces 2-12 mm
Arsenic
Formel: As
Molecular Weight: 74.92 g/mol
Schmelzpunkt: 817
Dichte: 5.727 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00085309
CAS-Nummer: 7440-38-2
EINECS: 231-148-6
UN: 1558
ADR: 6.1 II
Merck Index: 12,00832

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR