ω-Brom-3-chloracetophenon

Lieferant: BIONET RESEARCH

Synonyme: 2-Brom-3'-chloracetophenon, 3-Chlorphenacylbromid, α-Brom-3-chloracetophenon, Brommethyl-m-chlorphenylketon

10X-0707.5 10X-0707.500 10X-0707.10 10X-0707.1
BIOT10X-0707.5EA 0 EUR
BIOT10X-0707.5 BIOT10X-0707.500 BIOT10X-0707.10 BIOT10X-0707.1
ω-Brom-3-chloracetophenon
ω-Brom-3-chloracetophenon
Formel: C₈H₆BrClO
Molecular Weight: 233.49 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00792669
CAS-Nummer: 41011-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR