3-Bromimidazo[1,2-a]pyridin

Lieferant: BIONET RESEARCH

10X-0880.5 10X-0880.1 10X-0880.10 10X-0880.500
BIOT10X-0880.5EA 0 EUR
BIOT10X-0880.5 BIOT10X-0880.1 BIOT10X-0880.10 BIOT10X-0880.500
3-Bromimidazo[1,2-a]pyridin
3-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
MDL: MFCD00014556
CAS-Nummer: 4926-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR