1-(4-Nitrophenyl)-4-piperidincarbonsäure

Lieferant: BIONET RESEARCH

11H-951.500 11H-951.1 11H-951.5 11H-951.10
BIOT11H-951.500EA 0 EUR
BIOT11H-951.500 BIOT11H-951.1 BIOT11H-951.5 BIOT11H-951.10
1-(4-Nitrophenyl)-4-piperidincarbonsäure
1-(4-Nitrophenyl)-4-piperidincarbonsäure
Formel: C₁₂H₁₄N₂O₄
CAS-Nummer: 223786-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR