1-(4-Nitrophenyl)-4-piperidincarbonsäure
Lieferant: BIONET RESEARCH
11H-951.500
11H-951.1
11H-951.5
11H-951.10
BIOT11H-951.500EA
0
EUR
BIOT11H-951.500
BIOT11H-951.1
BIOT11H-951.5
BIOT11H-951.10
1-(4-Nitrophenyl)-4-piperidincarbonsäure
1-(4-Nitrophenyl)-4-piperidincarbonsäure
Formel:
C₁₂H₁₄N₂O₄ |
CAS-Nummer:
223786-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...