3,5-Dibrom-1H-1,2,4-triazol

Lieferant: BIONET RESEARCH

Synonyme: 3,5-Dibromo-1,2,4-triazole

11N-725.500 11N-725.1 11N-725.5 11N-725.10
BIOT11N-725.500EA 0 EUR
BIOT11N-725.500 BIOT11N-725.1 BIOT11N-725.5 BIOT11N-725.10
3,5-Dibrom-1H-1,2,4-triazol
3,5-Dibrom-1H-1,2,4-triazol
Formel: C₂HBr₂N₃
Molecular Weight: 226.8600 g/mol
MDL: MFCD00487260
CAS-Nummer: 7411-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR