3,5-Dibrom-1H-1,2,4-triazol
Lieferant: BIONET RESEARCH
Synonyme:
3,5-Dibromo-1,2,4-triazole
11N-725.500
11N-725.1
11N-725.5
11N-725.10
BIOT11N-725.500EA
0
EUR
BIOT11N-725.500
BIOT11N-725.1
BIOT11N-725.5
BIOT11N-725.10
3,5-Dibrom-1H-1,2,4-triazol
3,5-Dibrom-1H-1,2,4-triazol
Formel:
C₂HBr₂N₃ Molecular Weight: 226.8600 g/mol |
MDL:
MFCD00487260 CAS-Nummer: 7411-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...