6-Morpholino-3-pyridinamin

Lieferant: BIONET RESEARCH

Synonyme: 6-Morpholinopyridin-3-amine, 5-Amino-2-(4-morpholinyl)pyridine

11R-0626.500 11R-0626.1 11R-0626.5
BIOT11R-0626.500EA 0 EUR
BIOT11R-0626.500 BIOT11R-0626.1 BIOT11R-0626.5
6-Morpholino-3-pyridinamin
6-Morpholino-3-pyridinamin
Formel: C₉H₁₃N₃O
Molecular Weight: 179.22 g/mol
MDL: MFCD00119562
CAS-Nummer: 52023-68-4
EINECS: 257-613-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR