2-Brom-4'-(trifluormethyl)acetophenon

Lieferant: BIONET RESEARCH

Synonyme: 4-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one

11X-0702.500 11X-0702.1 11X-0702.5 11X-0702.10
BIOT11X-0702.500EA 0 EUR
BIOT11X-0702.500 BIOT11X-0702.1 BIOT11X-0702.5 BIOT11X-0702.10
2-Brom-4'-(trifluormethyl)acetophenon
2-Brom-4'-(trifluormethyl)acetophenon
Formel: C₉H₆BrF₃O
Molecular Weight: 267.0400 g/mol
MDL: MFCD00126489
CAS-Nummer: 383-53-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR