2-(4-Chlorphenyl)-4-thiazolcarbonsäure

Lieferant: BIONET RESEARCH

12F-339S.500 12F-339S.1 12F-339S.5 12F-339S.10
BIOT12F-339S.500EA 0 EUR
BIOT12F-339S.500 BIOT12F-339S.1 BIOT12F-339S.5 BIOT12F-339S.10
2-(4-Chlorphenyl)-4-thiazolcarbonsäure
2-(4-Chlorophenyl)-4-thiazolecarboxylic acid
Formel: C₁₀H₆NO₂ClS
MDL: MFCD00142032
CAS-Nummer: 17228-98-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR