2-(4-Bromphenylsulfonamido)essigsäure
Lieferant: BIONET RESEARCH
12G-419S.500
12G-419S.1
12G-419S.5
12G-419S.10
BIOT12G-419S.500EA
0
EUR
BIOT12G-419S.500
BIOT12G-419S.1
BIOT12G-419S.5
BIOT12G-419S.10
2-(4-Bromphenylsulfonamido)essigsäure
2-(4-Bromphenylsulfonamido)essigsäure
Formel:
C₈H₈BrNO₄S |
MDL:
MFCD01935943 CAS-Nummer: 13029-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...