2-(4-Bromphenylsulfonamido)essigsäure

Lieferant: BIONET RESEARCH

12G-419S.500 12G-419S.1 12G-419S.5 12G-419S.10
BIOT12G-419S.500EA 0 EUR
BIOT12G-419S.500 BIOT12G-419S.1 BIOT12G-419S.5 BIOT12G-419S.10
2-(4-Bromphenylsulfonamido)essigsäure
2-(4-Bromphenylsulfonamido)essigsäure
Formel: C₈H₈BrNO₄S
MDL: MFCD01935943
CAS-Nummer: 13029-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR