ω-Brom-3-cyanacetophenon

Lieferant: BIONET RESEARCH

Synonyme: 3-Cyanphenacylbromid, α-Brom-3-cyanacetophenon, 2-Brom-3'-cyanacetophenon, 3-(Bromacetyl)benzonitril, Brommethyl-m-cyanphenylketon

12X-0924.500 12X-0924.1 12X-0924.10 12X-0924.5
BIOT12X-0924.500EA 0 EUR
BIOT12X-0924.500 BIOT12X-0924.1 BIOT12X-0924.10 BIOT12X-0924.5
ω-Brom-3-cyanacetophenon
ω-Brom-3-cyanacetophenon
Formel: C₉H₆BrNO
MDL: MFCD00833267
CAS-Nummer: 50916-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR