2-Brom-N-(2,6-dimethylphenyl)acetamid

Lieferant: BIONET RESEARCH

1G-022.500 1G-022.1 1G-022.5 1G-022.10
BIOT1G-022.500EA 0 EUR
BIOT1G-022.500 BIOT1G-022.1 BIOT1G-022.5 BIOT1G-022.10
2-Brom-N-(2,6-dimethylphenyl)acetamid
2-Brom-N-(2,6-dimethylphenyl)acetamid
Formel: C₁₀H₁₂BrNO
Molecular Weight: 242.12 g/mol
MDL: MFCD00215011
CAS-Nummer: 32433-61-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR