2-(4-Pyridinyl)-4-thiazolcarbonsäure

Lieferant: BIONET RESEARCH

1G-331S.500 1G-331S.1 1G-331S.5 1G-331S.10
BIOT1G-331S.500EA 0 EUR
BIOT1G-331S.500 BIOT1G-331S.1 BIOT1G-331S.5 BIOT1G-331S.10
2-(4-Pyridinyl)-4-thiazolcarbonsäure
2-(4-Pyridinyl)-4-thiazolcarbonsäure
Formel: C₉H₆N₂O₂S
Molecular Weight: 206.22 g/mol
MDL: MFCD00171745
CAS-Nummer: 21278-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR