2-Ethyl-5-nitrobenzoxazol

Lieferant: BIONET RESEARCH

1W-0930.500 1W-0930.1 1W-0930.5 1W-0930.10
BIOT1W-0930.500EA 0 EUR
BIOT1W-0930.500 BIOT1W-0930.1 BIOT1W-0930.5 BIOT1W-0930.10
2-Ethyl-5-nitrobenzoxazol
2-Ethyl-5-nitrobenzoxazole
Formel: C₉H₈N₂O₃
CAS-Nummer: 204771-74-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR