Pentafluorphenyl-2,4-dichlorbenzolsulfonat

Lieferant: BIONET RESEARCH

1Y-0271.1 1Y-0271.10 1Y-0271.5 1Y-0271.500
BIOT1Y-0271.1EA 0 EUR
BIOT1Y-0271.1 BIOT1Y-0271.10 BIOT1Y-0271.5 BIOT1Y-0271.500
Pentafluorphenyl-2,4-dichlorbenzolsulfonat
Pentafluorphenyl-2,4-dichlorbenzolsulfonat
Formel: C₁₂H₃Cl₂F₅O₃S
MDL: MFCD06659543
CAS-Nummer: 885949-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR