ω-Brom-2-chloracetophenon

Lieferant: BIONET RESEARCH

1Y-0714.1 1Y-0714.10 1Y-0714.500 1Y-0714.5
BIOT1Y-0714.1EA 0 EUR
BIOT1Y-0714.1 BIOT1Y-0714.10 BIOT1Y-0714.500 BIOT1Y-0714.5
ω-Brom-2-chloracetophenon
ω-Brom-2-chloracetophenon
Formel: C₈H₆BrClO
Lagertemperatur: Kühlschrank
MDL: MFCD00832993
CAS-Nummer: 5000-66-8
UN: 1760
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR