2-Chlorisonicotinonitril
Lieferant: BIONET RESEARCH
1Y-0715.1
1Y-0715.10
1Y-0715.5
1Y-0715.500
BIOT1Y-0715.1EA
0
EUR
BIOT1Y-0715.1
BIOT1Y-0715.10
BIOT1Y-0715.5
BIOT1Y-0715.500
2-Chlorisonicotinonitril
2-Chlorisonicotinonitril
Formel:
C₆H₃ClN₂ |
MDL:
MFCD00174318 CAS-Nummer: 33252-30-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...