2-Amino-5-(4-fluorphenyl)-4-thiazolcarbohydrazid

Lieferant: BIONET RESEARCH

1Y-0931.1 1Y-0931.10 1Y-0931.5 1Y-0931.500
BIOT1Y-0931.1EA 0 EUR
BIOT1Y-0931.1 BIOT1Y-0931.10 BIOT1Y-0931.5 BIOT1Y-0931.500
2-Amino-5-(4-fluorphenyl)-4-thiazolcarbohydrazid
2-Amino-5-(4-fluorphenyl)-4-thiazolcarbohydrazid
Formel: C₁₀H₉FN₄OS
MDL: MFCD06200939
CAS-Nummer: 886361-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR