1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd

Lieferant: BIONET RESEARCH

2H-989.500 2H-989.1 2H-989.5 2H-989.10
BIOT2H-989.500EA 0 EUR
BIOT2H-989.500 BIOT2H-989.1 BIOT2H-989.5 BIOT2H-989.10
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd
Formel: C₁₅H₈ClF₃N₂O
MDL: MFCD00664496
CAS-Nummer: 338770-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR