(2-Chlor-3-pyridinyl)methanol
Lieferant: BIONET RESEARCH
2L-375S.500
2L-375S.1
2L-375S.5
2L-375S.10
BIOT2L-375S.500EA
0
EUR
BIOT2L-375S.500
BIOT2L-375S.1
BIOT2L-375S.5
BIOT2L-375S.10
(2-Chlor-3-pyridinyl)methanol
(2-Chlor-3-pyridinyl)methanol
Formel:
C₆H₆ClNO |
MDL:
MFCD01935961 CAS-Nummer: 42330-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...