4'-Chlorbiphenyl-4-carbonsäure
Lieferant: BIONET RESEARCH
Synonyme:
4'-Chloro[1,1'-biphenyl]-4-carboxylic acid, 4'-Chloro-[1,1'-biphenyl]-4-carboxylic acid
2X-0702.500
2X-0702.1
2X-0702.5
2X-0702.10
BIOT2X-0702.500EA
0
EUR
BIOT2X-0702.500
BIOT2X-0702.1
BIOT2X-0702.5
BIOT2X-0702.10
4'-Chlorbiphenyl-4-carbonsäure
4'-Chlorbiphenyl-4-carbonsäure
Formel:
C₁₃H₉ClO₂ |
MDL:
MFCD03424592 CAS-Nummer: 5748-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...