Methyl-4'-nitrobiphenyl-4-carboxylat
Lieferant: BIONET RESEARCH
Synonyme:
Methyl 4'-nitro[1,1'-biphenyl]-4-carboxylate
2X-0709.500
2X-0709.1
2X-0709.5
2X-0709.10
BIOT2X-0709.500EA
0
EUR
BIOT2X-0709.500
BIOT2X-0709.1
BIOT2X-0709.5
BIOT2X-0709.10
Methyl-4'-nitrobiphenyl-4-carboxylat
Methyl-4'-nitrobiphenyl-4-carboxylat
Formel:
C₁₄H₁₁NO₄ Molecular Weight: 257.25 g/mol |
MDL:
MFCD04117797 CAS-Nummer: 5730-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...