Methyl-4'-nitrobiphenyl-4-carboxylat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 4'-nitro[1,1'-biphenyl]-4-carboxylate

2X-0709.500 2X-0709.1 2X-0709.5 2X-0709.10
BIOT2X-0709.500EA 0 EUR
BIOT2X-0709.500 BIOT2X-0709.1 BIOT2X-0709.5 BIOT2X-0709.10
Methyl-4'-nitrobiphenyl-4-carboxylat
Methyl-4'-nitrobiphenyl-4-carboxylat
Formel: C₁₄H₁₁NO₄
Molecular Weight: 257.25 g/mol
MDL: MFCD04117797
CAS-Nummer: 5730-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR