2-Hydrazino-3-nitropyridin

Lieferant: BIONET RESEARCH

3F-306S.500 3F-306S.1 3F-306S.5 3F-306S.10
BIOT3F-306S.500EA 0 EUR
BIOT3F-306S.500 BIOT3F-306S.1 BIOT3F-306S.5 BIOT3F-306S.10
2-Hydrazino-3-nitropyridin
2-Hydrazino-3-nitropyridin
Formel: C₅H₆N₄O₂
MDL: MFCD00085923
CAS-Nummer: 15367-16-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR